Lock Out/Tag Out; Danger - Do Not Operate (white block) - Equipment Locked Out By
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
£16.41GBP / Pkg of 25
£19.69GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
Accuform MLT409CTP Lockout/Tagout Tag, Danger-Do Not Operate, Block, 5.8x3.1", Cardstock; 25/Pk WZ-40503-03 Mfr # MLT409CTP
COMPARE
Polyolefin
£16.41GBP / Pkg of 25
£19.69GBP (incl VAT)