This product is now discontinued, View Related Products
Laxco Analog Salt Refractometers
Ideal for aquaculture, food processing, and wildlife management
  • Choose the meter range, in percent or ppt, to suit your application needs
Measure salt content in parts per thousand (ppt) and specific gravity, or percent salinity (depending on mode.)
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Salinity Range (%) Resolution
Availability Pricing
Laxco RHS-10ATC Low-cost salt refractometer, 0 to 100 ppt salinity WZ-81150-20 Mfr # RHS-10ATC
COMPARE
1 (ppt); 0.001 (s.g.)
Call For Price
Laxco RHS-28 Low-cost salt refractometer, 0 to 28% salinity WZ-81150-22 Mfr # RHS-28
COMPARE
28 0.2% (salinity)