This product is now discontinued, View Related Products
Lascar Electronics Refrigerator and Freezer Data Loggers
Three models to choose from for compact, Wi-Fi enabled, or dual channel monitoring
  • All loggers feature high-accuracy thermistor temperature probes in a glycol-filled bottle — perfect for refrigerator and freezer temperature monitoring
  • NIST-traceable calibration certificate included with each logger to assure accuracy to ±0.1°C (±0.18°F) from -10 to 70°C (14 to 158°F)
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lascar Electronics Compact USB Data Logger with Thermistor in Glycol-Filled Bottle WZ-18000-67 Mfr # EL-USB-TP-LCD-PROBE-G
COMPARE
Compact USB Data Logger with Thermistor in Glycol-Filled Bottle
Call For Price
Lascar Electronics Wi-Fi Enabled Data Logger with Thermistor Probe in Glycol-Filled Bottle WZ-18000-77 Mfr # EL-WIFI-TP+ -PROBE-G
COMPARE
Wi-Fi Enabled Data Logger with Thermistor Probe in Glycol-Filled Bottle
Call For Price
Lascar Electronics Dual Channel Data Logger with Thermistor Probe in Glycol-Filled Bottle WZ-18000-87 Mfr # EL-GFX-DTP-PROBE-G
COMPARE
Dual Channel Data Logger with Thermistor Probe in Glycol-Filled Bottle