Lascar Electronics EL-GFX USB Data Loggers with Real-time Data Analysis
Graphic LCD provides real-time information on data being recorded
  • Ability to start, stop, graph trends, and summarize data in the field without computer connection
  • Memory capacity exceeds 250,000 readings per channel with resolution to the nearest 0.1 digit
  • Windows®-based control software available as free download
  • Compact and robust waterproof design with IP67 rating
  • Wall-mounting clip for quick magnetic mounting to a metallic surface
£155.91 - £230.00GBP / Each
£187.09 - £276.00GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Lascar Electronics EL-GFX-1 Temperature Data Logger with Graphic LCD Screen WZ-23039-04 Mfr # EL-GFX-1
COMPARE
176 ±0.2 0.1
£155.91GBP / Each
£187.09GBP (incl VAT)
Lascar Electronics EL-GFX-2 Temperature and Humidity Data Logger with Graphic LCD Screen WZ-23039-05 Mfr # EL-GFX-2
COMPARE
176 ±1.0 0.1
£172.28GBP / Each
£206.74GBP (incl VAT)
Lascar Electronics EL-GFX-DTC Dual Channel Thermocouple Data Logger with Graphic LCD Screen WZ-23039-06 Mfr # EL-GFX-DTC
COMPARE
±0.9 0.1 to 999.9, 1 above 999.9
£213.19GBP / Each
£255.83GBP (incl VAT)
Lascar Electronics EL-GFX-TC USB Data Logger with Graphic LCD Screen, single thermocouple WZ-23039-11 Mfr # EL-GFX-TC
COMPARE
£178.64GBP / Each
£214.37GBP (incl VAT)
Lascar Electronics EL-GFX-DTP USB Data Logger with Graphic LCD Screen, dual thermistor WZ-23039-12 Mfr # EL-GFX-DTP
COMPARE
257
£213.19GBP / Each
£255.83GBP (incl VAT)
Lascar Electronics EL-GFX-2+ Temperature and Humidity Data Logger with Graphic LCD Screen WZ-23039-14 Mfr # EL-GFX-2+
COMPARE
257
£230.00GBP / Each
£276.00GBP (incl VAT)