This product is now discontinued, View Related Products
Large-Diameter Circular Illuminated Magnifiers
High-quality glass provides clear and aberration-free magnification
Large 7-1/2" diameter lens and full-spectrum fluorescent bulb provide excellent viewing. Heavy-duty construction includes 1/2" square arms with smooth spring-loaded movement, cast-iron clamp, and ESD-safe shade. The 10" (25.4 cm) dia. circular, 34-watt bulb provides shadowless illumination. Swivel mount allows sideways and angular mounting for nearly limitless positioning. Magnifiers operate on 120 VAC.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Arm Length (in) Magnification Viewing Area
Availability Pricing
O C White 52400 Large-diameter Circular Illuminated Magnifier, 1.75x magnification WZ-41807-80 Mfr # 52400
COMPARE
43 1.75x 7.5\" (190.5 mm) diameter
Call For Price
O C White 52400-4 Large-diameter Circular Illuminated Magnifier, 2x magnification WZ-41807-85 Mfr # 52400-4
COMPARE
43 2x 7.5\" (190.5 mm) diameter
Call For Price
O C White 52400-4 Large-diameter Circular Illuminated Magnifier, 2.25x magnification WZ-41807-86 Mfr # 52400-5
COMPARE
43 2.25x 7.5\" (190.5 mm) diameter
Call For Price
O C White 51600 Large-diameter Circular Illuminated Magnifier, 1.75x magnification, 25" reach WZ-41807-91 Mfr # 51600
COMPARE
25 1.75x 7.5\" (190.5 mm) diameter