Cole-Parmer® TELOS® Florisil® Polar SPE Chromatography Columns
Extract polar analytes from nonpolar organic solvents
  • Commonly used for separation of lipids and pesticide residue extraction
  • Compatible with manual and automated sample processing protocols
Use the TELOS polar (normal phase) SPE columns to retain the analytes of interest or to remove polar interferences from the sample. Columns with Florisil® PR grade (magnesium silicate) sorbent have larger particle size than silica and NH2. Florisil has a pore diameter of 70Å. All columns are made of polypropylene (except 06476-29, made of glass) and feature Luer-slip outlet.
£97.71 - £162.85GBP / Pkg of 30
£117.25 - £195.42GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg) Wetted Materials
Availability Pricing
COMPARE
6 1000 Polypropylene body and polyethylene frits
£113.99GBP / Pkg of 30
£136.79GBP (incl VAT)
COMPARE
6 1000 Glass body and polyethylene frits
£162.85GBP / Pkg of 30
£195.42GBP (incl VAT)
COMPARE
15 2000 Polypropylene body and polyethylene frits
£113.99GBP / Pkg of 20
£136.79GBP (incl VAT)
COMPARE
3 500 Polypropylene body and polyethylene frits
Call For Price
COMPARE
6 500 Polypropylene body and polyethylene frits
£97.71GBP / Pkg of 30
£117.25GBP (incl VAT)