Heraeus Noblelight Hollow Cathode Lamps, Vanadium
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
Heraeus Noblelight vanadium (V) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.
£315.41 - £409.13GBP / Pkg of 1
£378.49 - £490.96GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
Vanadium Uncoded Hollow Cathode Lamp, vanadium, 37 mm, uncoded
£315.41GBP / Pkg of 1
£378.49GBP (incl VAT)
COMPARE
Vanadium Unicam Hollow Cathode Lamp, vanadium, 37 mm, Unicam coded
£352.08GBP / Pkg of 1
£422.50GBP (incl VAT)
COMPARE
Vanadium Varian Hollow Cathode Lamp, vanadium, 37 mm, Varian coded
£352.08GBP / Pkg of 1
£422.50GBP (incl VAT)
COMPARE
Vanadium Perkin Elmer Lumina Hollow Cathode Lamp, vanadium, 50 mm, Perkin Elmer Lumina coded
£409.13GBP / Pkg of 1
£490.96GBP (incl VAT)