Kimcote™ Safety Coated Class A Volumetric Flasks with Glass Stopper
KimCote™ plastic coating for added safety
  • Reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Volumetric flasks are constructed of low extractable borosilicate glass
Volumetric plastic-coated flasks feature white ceramic enamel marking spots and glass stoppers. Calibrated "To Contain", flasks include a single graduation ring. Flasks are designed to meet ASTM Spec E288, Class A unserialized requirements. Autoclavable to 120°C (250°F).
£420.44 - £1,051.74GBP / Case of 12
£504.53 - £1,262.09GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stopper Number Accepted Tolerance (mL)
Availability Pricing
DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 100 mL WZ-34524-31 Mfr # KC28014-100
COMPARE
100 13 ±0.08
£1,051.74GBP / Case of 12
£1,262.09GBP (incl VAT)
DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 1000 mL WZ-34524-37 Mfr # KC28014-1000
COMPARE
1000 22 ±0.3
£661.24GBP / Case of 4
£793.49GBP (incl VAT)
DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 2000 mL WZ-34524-39 Mfr # KC28014-2000
COMPARE
2000 27 ±0.5
£420.44GBP / Case of 2
£504.53GBP (incl VAT)