Kimble® CHROMAFLEX® Flow Adapters for Standard and Jacketed Columns
Optimixe performance of your CHROMAFLEX® columns
  • Recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
Improve chromatogram resolution by delivering the sample and buffer directly to top of gel bed. This protects gel bed from disruption during sample loading and eliminates any dead volume above gel bed. They are ideal for concentration and pH gradients, reverse flow separations, and high-resolution gravity flow. Each includes a 20 µm porosity HDPE bed support, PTFE/propylene O-ring seal, stainless steel adjusting rod, PTFE body and five feet (1.5 m) of FEP PTFE tubing.
£339.44 - £780.59GBP / Case of 1
£407.33 - £936.71GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in) Description
Availability Pricing
DWK Life Sciences (Kimble) CHROMAFLEX® Flow Adapter for Standard Columns, 1.0 cm ID x 1/16" OD Tubing WZ-34696-54 Mfr # 420836-0000
COMPARE
10 0.002 Flow adapter, for 1.0 cm ID standard columns
£339.44GBP / Case of 1
£407.33GBP (incl VAT)
DWK Life Sciences (Kimble) CHROMAFLEX® Flow Adapter for Jacketed Columns, 2.5 cm IID x 1/8" OD Tubing WZ-34696-66 Mfr # 420876-0020
COMPARE
25 0.002 Flow adapter, for 2.5 cm ID jacketed columns, 1/8\" OD tubing
£549.91GBP / Case of 1
£659.89GBP (incl VAT)
DWK Life Sciences (Kimble) CHROMAFLEX® Flow Adapter for Jacketed Columns, 4.8 cm ID x 1/8" OD Tubing WZ-34696-68 Mfr # 420876-0040
COMPARE
48 0.005 Flow adapter, for 4.8 cm ID jacketed columns
£780.59GBP / Case of 1
£936.71GBP (incl VAT)