Kimax Glass Dropping Bottles
Clear borosilicate glass bottles conform to ASTM D 281 requirements for use in determining the oil absorption of pigments. Bottles 34523–32 and -34 meet Federal NNN-B-1194, Type II, Class 2, Grade B standards. Pipette style 1-mL glass dropper has a rubber nipple.
£666.45 - £1,123.77GBP / Case of 12
£799.74 - £1,348.52GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
DWK Life Sciences (Kimble) 15035-15 Glass Dropper Bottle, 15 mL, 12/Cs WZ-34523-30 Mfr # 15035-15
COMPARE
15 12/18 Glass
£666.45GBP / Case of 12
£799.74GBP (incl VAT)
DWK Life Sciences (Kimble) 15035-60 Glass Dropper Bottle, 60 mL, 12/Cs WZ-34523-34 Mfr # 15035-60
COMPARE
60 18/18 Glass
£829.59GBP / Case of 12
£995.51GBP (incl VAT)
DWK Life Sciences (Kimble) 15035-125 Glass Dropper Bottle, 125 mL, 12/Cs WZ-34523-36 Mfr # 15035-125
COMPARE
125 19/22 Glass
£1,123.77GBP / Case of 12
£1,348.52GBP (incl VAT)