Keck Glassware Quick Disconnect Adapters
Safely and easily connect tubing to glassware or other tubing
Model 06841-51 glassware adapter allows tubing adapter to fit GL-14 threaded port (Schott glass) or 10-mm OD barbed port. Adapter 06841-52 allows tubing adapter to fit RD-14 threaded port (Normag glass) or 11-mm OD barbed port. These glassware adapters are suitable for use with tubing adapters 06841-50 and -60, sold separatley.
£93.85GBP / Pkg of 10
£112.62GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Keck Glassware Adapter, Gl-14 Threaded Port or 10 mm OD Barbed Port, 10/Pk WZ-06841-51 Mfr # MAKE-UP
COMPARE
Glassware Adapter, Gl-14 Threaded Port or 10 mm OD Barbed Port, 10/Pk
£93.85GBP / Pkg of 10
£112.62GBP (incl VAT)