Justrite Wooden Laminate Storage Cabinets
Ideal for storing aggressive corrosives
  • Shelf adjusts on 1 1/4" (32-mm) centers
  • Easy-to-clean polyethylene work surface
Long-lasting wooden laminate storage cabinet with stainless steel handles and hinges provides exceptional strength. Shelf adjusts on 1-1/4-in (32-mm) centers. Cabinet includes adjustable leg levelers, Haz-Alert™ reflective labels, and double key set for safety and security. A removable, easy-to-clean polyethylene work surface is recessed 5/8-in (16-mm) to contain leaks.
£851.24GBP / Each
£1,021.49GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Color Height (in)
Availability Pricing
Justrite 24120 Wooden Laminate Corrosive Safety Cabinet, Benchtop, Manual Close, 2 Doors, 4 Gal WZ-86447-33 Mfr # 24120
COMPARE
4 Blue 15 3/4
£851.24GBP / Each
£1,021.49GBP (incl VAT)