Justrite® Quick-Disconnect Disposal Can
Remove can and empty waste without disconnecting your inlet tubing
  • Safe method to stage and draw reagents
  • Quick disconnects allow simple attachment of one inlet and one vent
  • Eliminate safety concerns and accidents associated with glass bottles
  • FM approved
  • EPA complaint
Quick-disconnect safety cans provide a safe hands-free way to drain waste from your HPLC application. Prevent the escape of ignitable and toxic vapors. Protect the liquid and vapors inside from external heat sources. The quick disconnect mechanisms are valved open when connected and valved closed when disconnected for spill free changeout of safety cans without disconnecting the tubing.
Coalescing carbon filter attaches to one of the fittings to filter VOCs. Durable polyethylene is translucent allowing you quick visual inspection of liquid level. Pouring spout includes stainless steel flame arrester to prevent flashback ignition. Dual fittings eliminate back pressure and provide controlled handling of vapors. Container automatically vents at 3-5 psig.
An optional spill basin (86394-45) captures any leakage and provides additional protection against accidental pumping.
£323.11 - £1,880.46GBP / Each
£387.73 - £2,256.55GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Capacity (Liters)
Availability Pricing
Justrite 12160 Quick-Disconnect Disposal Can, Polypropylene Fitting, 1 Gal WZ-12021-08 Mfr # 12160
COMPARE
1 4
£323.11GBP / Each
£387.73GBP (incl VAT)
Justrite 12165 Quick-Disconnect Disposal Can, Stainless Steel or Polypropylene Fittings, 2 Gal WZ-12021-09 Mfr # 12165
COMPARE
2 8
£1,023.10GBP / Each
£1,227.72GBP (incl VAT)
Justrite Quick-Disconnect Disposal Can, Stainless Steel Fittings; 2 Gal WZ-12030-72 Mfr # TF12752
COMPARE
2 8
£1,880.46GBP / Each
£2,256.55GBP (incl VAT)