Justrite Liquid Disposal Safety Cans, Polyethylene
Allows quick visual inspection of liquid level
  • Flame arrester guards against outside ignition
  • Automatically vents at between 3 and 5 psig
  • OSHA and NFPA compliant
  • FM Approved
These liquid disposal safety cans offer you a better solution for the containment of flammable liquid or flammable liquid waste. Constructed of durable translucent polyethylene makes it easily to detect liquid level. Large opening with cap held open provides convenient filling and pouring. The cans also offer superior chemical resistance and leak protection.
Conductive, patented carbon insert embedded in the container allows for proper grounding. Broad-base design provides stability and resists accidental tip over. Self-closing lid automatically vents between 3 and 5 psi (0.2 and 0.35 bar). Built-in stainless steel flame arrester guards against inside vapor ignition. Stainless steel hardware is corrosive resistant.
£227.71 - £388.38GBP / Each
£273.25 - £466.06GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Capacity (Liters)
Availability Pricing
Justrite 12751 Liquid Disposal Safety Can, Polyethylene, 2 Gal WZ-86395-20 Mfr # 12751
COMPARE
2 7.57
£227.71GBP / Each
£273.25GBP (incl VAT)
Justrite 12754 Liquid Disposal Safety Can, Polyethylene, 5 Gal WZ-86395-24 Mfr # 12754
COMPARE
5 18.93
£388.38GBP / Each
£466.06GBP (incl VAT)