This product is now discontinued, View Related Products

Julabo Viscocity Baths

Save time by holding multiple viscometers at one time
  • Hold multiple viscometers at a time — ideal for calibration or comparison procedures
  • Low-level audible and optical alarms protect your samples and the unit
  • Highest accuracy with 3-point calibration and PID temperature control
A wide variety of temperature ranges make these viscometer baths ideal for laboratory and general-use applications. All baths feature a VFD Comfort display for simultaneously displaying three values, warning functions, high temperature cut-off, and pump stages. Easily transfer data to a PC via the RS-232 interface. The smart pump technology offers consistent pump performance with the electronically adjustable pump stages. Ensure maximum safety with adjustable high-temperature cutoff, dry running protection, and low liquid level warnings with both optical and audible alarms. The PID3 temperature control is ideal for higher demands with a temperature stability of ±0.01°C. Maximum accuracy is achieved with the Pt100 external temperature sensor for directly measuring and controlling external applications. Easy program control of temperature and time is achieved using one temperature profile with 10 steps.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description Power (VAC)
Availability Pricing
Julabo ME-31A 31L Viscometer Bath; 100-115V/50-60Hz WZ-12150-80 Mfr # 9162331.22
COMPARE
31L Viscometer Bath, 100-115V/50-60Hz 115
Call For Price
Julabo ME-31A 31L Viscometer Bath; 230V/50-60Hz WZ-12150-81 Mfr # 9162331.33
COMPARE
31L Viscometer Bath, 230V/50-60Hz 230
Call For Price
Julabo ME-18V 18L Viscometer Bath, 100-115V/50-60Hz SKU 1215082 WZ-12150-82 Mfr # 9162518.22
COMPARE
18L Viscometer Bath, 100-115V/50-60Hz 115
Call For Price
Julabo ME-18V 18L Viscometer Bath, 230V/50-60Hz SKU 1215083 WZ-12150-83 Mfr # 9162518.33
COMPARE
18L Viscometer Bath, 230V/50-60Hz 230
Call For Price
Julabo ME-18V 18L Viscometer Bath, 100-115V/50-60Hz SKU 1215084 WZ-12150-84 Mfr # 9162518.22.TT
COMPARE
18L Viscometer Bath, 100-115V/50-60Hz 115
Call For Price
Julabo ME-18V 18L Viscometer Bath, 230V/50-60Hz SKU 1215085 WZ-12150-85 Mfr # 9162518.33.TT
COMPARE
18L Viscometer Bath, 230V/50-60Hz 230
Call For Price
Julabo ME-16G 16L Viscometer Bath; 100-115V/50-60Hz WZ-12150-86 Mfr # 9162616.22
COMPARE
16L Viscometer Bath, 100-115V/50-60Hz 115
Call For Price
Julabo ME-16G 16L Viscometer Bath; 230V/50-60Hz WZ-12150-87 Mfr # 9162616.33
COMPARE
16L Viscometer Bath, 230V/50-60Hz 230
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat