This product is now discontinued, View Related Products
Julabo HighTech Heating Circulators, Six Programs
Highly stable temperature control improves system performance
  • Ensure repeatable results with ±0.01°C stability
  • Durable stainless-steel tanks withstand higher temperatures
  • RS-232 interface for optional remote control
  • Integrated cooling coil connects with tap water supply
  • Ideal for both internal and external circulation applications
These heating circulators offer superior temperature stability on all models and RS-232 interface for remote control. External pump connections and integrated cooling coils enhance the versatility of these circulating baths.
HighTech (HL, and SL series) circulators feature four-stage force and suction pumps for external closed loop and open system applications. The HL and SL series circulators provide expanded programming and data monitoring options with onboard memory for storage of up to 6 programs, and additional connections for supplementary pumps and valves.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Reservoir Capacity (L) Temperature Stability (° C)
Availability Pricing
Julabo HL-4 HighTech Programmable Heating Circulator, Six Programs, 4.5L; 100 to 115 VAC, 50/60 Hz WZ-12151-07 Mfr # 9312504.22
COMPARE
100 to 115 4.5 ±0.01
Call For Price
Julabo HL-4 HighTech Programmable Heating Circulator, Six Programs, 4.5L; 208 to 230 VAC, 50/60 Hz WZ-12151-08 Mfr # 9312504.33
COMPARE
208 to 230 4.5 ±0.01
Call For Price
Julabo SL-6 HighTech Programmable Heating Circulator, Six Programs, 6L; 208 to 230 VAC, 50/60 Hz WZ-12151-09 Mfr # 9352506.33
COMPARE
208 to 230 6 ±0.01
Call For Price
Julabo SL-12 HighTech Programmable Heating Circulator, Six Programs, 12L; 208 to 230 VAC, 50/60 Hz WZ-12151-10 Mfr # 9352512.33
COMPARE
208 to 230 12 ±0.01
Call For Price
Julabo SL-26 HighTech Programmable Heating Circulator, Six Programs, 26L; 208 to 230 VAC, 50/60 Hz WZ-12151-11 Mfr # 9352526.33
COMPARE
208 to 230 26 ±0.01