This product is now discontinued, View Related Products
Jonard Precision Dynamometer Tension Gauges
Measurement is extremely simple. Just place point of gauge arm perpendicular to the force to be measured. Reading can be taken in both directions- clockwise and counter-clockwise.
All Gauges Equipped With Maximum Reading Pointer
Maximum reading pointer moves with indicator when testing. It will remain at maximum force measured, enabling user to take reading afterward. No need to look at dial hand while measuring, thus avoiding reading errors.
Gauges Offer Extreme Precision And Durability
The measuring springs are made of hard-tempered annealed beryllium copper. This alloy provides outstanding flexibility with minimum effort. Dial is protected by unbreakable glass. Packaged in rigid, fitted plastic case to protect instrument.
All gauges equipped with maximum reading pointer…
(Reading + Maximum Scale Reading)
APPLICATIONS
  • Spring tension of all relay contacts and switches
  • Starting torque or motors, clock movements, etc.
  • Pressure of brushes in electro-mechanical devices, phono tone arms, needles, etc.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (g)
Availability Pricing
Jonard Industries GD-100 Gauge,Dynamometer 100-1000grams WZ-93955-07 Mfr # GD-100
COMPARE
1000
Call For Price
Jonard Industries GD-200 Gauge,Dynamometer 200-2000grams WZ-93955-09 Mfr # GD-200
COMPARE
2000
Call For Price
Jonard Industries GD-400 Gauge,Dynamometer 400-4000grams WZ-93955-12 Mfr # GD-400
COMPARE
4000
Call For Price
Jonard Industries GD-50 Precision Dynamometer; 60-500 g WZ-93955-14 Mfr # GD-50
COMPARE
500