John Guest polypropylene push-to-connect fittings
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.
£28.11 - £45.23GBP / Pkg of 10
£33.73 - £54.28GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Push-to-Connect Tube OD (port 2) (in)
Availability Pricing
John Guest Push-to-connect union elbows, polypropylene, 1/4", 10/pack WZ-34007-48 Mfr # PP0308W
COMPARE
Polypropylene 1/4 1/4
£31.13GBP / Pkg of 10
£37.36GBP (incl VAT)
John Guest Push-to-connect union elbows, polypropylene, 3/8", 10/pack WZ-34007-49 Mfr # PP0312W
COMPARE
Polypropylene 3/8 3/8
£45.23GBP / Pkg of 10
£54.28GBP (incl VAT)
John Guest Push-to-connect straight unions, polypropylene, 1/4", 10/pack WZ-34007-51 Mfr # PP0408W
COMPARE
Polypropylene 1/4 1/4
£28.11GBP / Pkg of 10
£33.73GBP (incl VAT)
John Guest Push-to-connect straight unions, polypropylene, 3/8", 10/pack WZ-34007-52 Mfr # PP0412W
COMPARE
Polypropylene 3/8 3/8
£37.87GBP / Pkg of 10
£45.44GBP (incl VAT)