John Guest acetal push-to-connect elbow adapters
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
£28.60 - £58.51GBP / Pkg of 10
£34.32 - £70.21GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Push-to-Connect Tube OD (port 1) (in) Push-to-Connect Tube OD (port 2) (in)
Availability Pricing
John Guest Push-to-connect stem elbow adapters, 1/4" x 1/4", 10/pack WZ-34006-76 Mfr # PI220808S
COMPARE
Acetal 1/4 1/4
£31.72GBP / Pkg of 10
£38.06GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 3/8" x 3/8", 10/pack WZ-34006-77 Mfr # PI221212S
COMPARE
Acetal 3/8 3/8
£48.85GBP / Pkg of 10
£58.62GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 1/2" x 1/2", 10/pack WZ-34006-78 Mfr # PI221616S
COMPARE
Acetal 1/2 1/2
£58.51GBP / Pkg of 10
£70.21GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 3/16" x 3/16", 10/pack WZ-34006-99 Mfr # PI220606S
COMPARE
Acetal 3/16 3/16
£28.60GBP / Pkg of 10
£34.32GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 3/8" x 1/4", 10/pack WZ-34007-02 Mfr # PI221208S
COMPARE
Acetal 3/8 1/4
£37.87GBP / Pkg of 10
£45.44GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 3/8" x 5/16", 10/pack WZ-34007-03 Mfr # PI221210S
COMPARE
Acetal 3/8 5/16
£37.87GBP / Pkg of 10
£45.44GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 5/32" x 5/32", 10/pack WZ-34007-35 Mfr # PM220404S
COMPARE
Acetal 5/32 5/32
£28.60GBP / Pkg of 10
£34.32GBP (incl VAT)
John Guest Push-to-connect stem elbow adapters, 5/16" x 5/16", 10/pack WZ-34007-36 Mfr # PM220808S
COMPARE
Acetal 5/16 5/16
£36.86GBP / Pkg of 10
£44.23GBP (incl VAT)
Showing 1- 8 of 8