Cole-Parmer® FF-200 Series Research Flame Photometer
Designed for research and analytical applications
  • Automatic flame failure detection for user safety
  • Built-in linearizer circuitry
  • Features sodium (Na), potassium (K), and lithium (Li) filters
The low temperature, single channel flame photometer is designed for routine determinations of sodium, potassium, and lithium. The unit features a flame failure safety system and a built-in linearizer circuitry which allows readings of both sodium and potassium to be displayed directly in mmol/liter.
£4,854.80GBP / Each
£5,825.76GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fuel Supply Power (VAC)
Availability Pricing
COMPARE
Propane, butane, natural gas and liquefied petroleum gas 230
£4,854.80GBP / Each
£5,825.76GBP (incl VAT)
COMPARE
Propane, butane, natural gas and liquefied petroleum gas 120