Ismatec® Standard Cartridge Pump Heads
Ideal pump-head for multichannel applications at higher flow rates
  • Snap your pump head onto your drives quickly with turn-and-lock mounting system
  • Select from sixteen interchangeable pump heads that offer flow rates from 0.01 to 3660 mL/min
  • RS-232 interface connects to your PC
Select from four-, eight-, or twelve-channel pump heads. They deliver a wide range of flow rates—as low as 0.01 mL/min to as high as 180 mL/min! Click ‘n’ go cartridges allow quick and easy tubing changes.
The first of a series of two-stop tubing options is below under Accessories. See all of our two-stop tubing formulations or call our Applications Specialists for personal assistance.
£822.59 - £1,585.18GBP / Each
£987.11 - £1,902.22GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Number of Channels Rollers
Availability Pricing
COMPARE
180 4 8
£822.59GBP / Each
£987.11GBP (incl VAT)
COMPARE
180 8 8
£1,274.20GBP / Each
£1,529.04GBP (incl VAT)
COMPARE
180 12 8
£1,585.18GBP / Each
£1,902.22GBP (incl VAT)