Ismatec® Sanitary Valveless Piston Pump Heads, Series Q
No valves to clog or wear out
  • Build your own pump systems using interchangeable heads, drives, and accessories
  • Pump fluids with viscosities up to 500 cp
  • Low dead volume
Choose ceramic materials for use with common abrasives. These sanitary high-flow models are ideal for pumping food products. Pump heads with ceramic piston and carbon cylinder liner are great for gas and liquid transfer.
£702.55GBP / Each
£843.06GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Max Flow Rate (Strokes/min) Max Pressure (PSI)
Availability Pricing
COMPARE
192 600 100
£702.55GBP / Each
£843.06GBP (incl VAT)
COMPARE
432 600 100
£702.55GBP / Each
£843.06GBP (incl VAT)