Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps
Independently control flow and direction of each channel via keypad or your PC
  • Flow rate 0.001 to 43 mL/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy.
Order the three-stop tubing size and formulation to suit your application needs (sold separately). More than 100 options exist — the most popular sizes and formulations can be found under Accessories below
£1,737.72 - £3,079.34GBP / Each
£2,085.26 - £3,695.21GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Rollers Number of Channels
Availability Pricing
COMPARE
35 8 2
£1,737.72GBP / Each
£2,085.26GBP (incl VAT)
COMPARE
35 8 3
£1,990.77GBP / Each
£2,388.92GBP (incl VAT)
COMPARE
35 8 4
£2,244.40GBP / Each
£2,693.28GBP (incl VAT)
COMPARE
24 12 4
£2,328.87GBP / Each
£2,794.64GBP (incl VAT)
COMPARE
43 6 2
£2,076.87GBP / Each
£2,492.24GBP (incl VAT)
COMPARE
35 8 2
Call For Price
COMPARE
24 12 2
£2,399.78GBP / Each
£2,879.74GBP (incl VAT)
COMPARE
43 6 3
£2,508.23GBP / Each
£3,009.88GBP (incl VAT)
COMPARE
35 8 3
£2,853.74GBP / Each
£3,424.49GBP (incl VAT)
COMPARE
24 12 3
£2,674.71GBP / Each
£3,209.65GBP (incl VAT)
COMPARE
43 6 4
£2,404.30GBP / Each
£2,885.16GBP (incl VAT)
COMPARE
35 8 4
£2,851.63GBP / Each
£3,421.96GBP (incl VAT)
COMPARE
24 12 4
£3,079.34GBP / Each
£3,695.21GBP (incl VAT)
Showing 1- 13 of 13