Ismatec® Quick-Couple, Single-Channel Pump Heads for 1.6 or 2.4 mm Wall Tubing
Snap your pump head onto your drives quickly with turn-and-lock mounting system
  • Select from interchangeable pump heads that offer flow rates from 0.01 to 3660 mL/min
Ensure optimum performance from your Ismatec® pumps.
£678.86 - £1,727.93GBP / Each
£814.63 - £2,073.52GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Rollers
Availability Pricing
COMPARE
3700 0.49 2
£888.52GBP / Each
£1,066.22GBP (incl VAT)
COMPARE
3100 3.6 2
£911.17GBP / Each
£1,093.40GBP (incl VAT)
COMPARE
2900 3.3 3
£909.84GBP / Each
£1,091.81GBP (incl VAT)
COMPARE
3400 0.45 3
Call For Price
COMPARE
2800 0.44 3
£678.86GBP / Each
£814.63GBP (incl VAT)
COMPARE
880 0.072 3
£796.49GBP / Each
£955.79GBP (incl VAT)
COMPARE
3600 0.41 3
£1,727.93GBP / Each
£2,073.52GBP (incl VAT)
Showing 1- 7 of 7