Ismatec® Pump Tubing, Tygon® LMT-55
Exceptional durability with long life
  • Crystal clear and flexible while also being long-lasting and crack-resistant
Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments
Sterilize by autoclave or ethylene oxide gas.
£38.44 - £221.03GBP / Pkg of 1
£46.13 - £265.24GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
1.6 4.8 1.6
£38.44GBP / Pkg of 1
£46.13GBP (incl VAT)
COMPARE
3.2 6.4 1.6
£80.24GBP / Pkg of 1
£96.29GBP (incl VAT)
COMPARE
4.8 8 1.6
£66.87GBP / Pkg of 1
£80.24GBP (incl VAT)
COMPARE
6.4 9.6 1.6
£73.83GBP / Pkg of 1
£88.60GBP (incl VAT)
COMPARE
9.5 12.7 1.6
£152.07GBP / Pkg of 1
£182.48GBP (incl VAT)
COMPARE
6.4 11.1 2.4
£156.68GBP / Pkg of 1
£188.02GBP (incl VAT)
COMPARE
9.5 16 3.2
£221.03GBP / Pkg of 1
£265.24GBP (incl VAT)
Showing 1- 7 of 7