Ismatec® Pump Tubing, 3-Stop Microbore, FDA-Compliant Viton®
Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Perfect for food and lab applications where FDA compliance is required
  • Excellent chemical compatibility
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
FDA-Compliant Viton® tubing features an ultra-smooth inner bore, excellent biocompatibility for sensitive applications, non-cytotoxic, non-pyrogenic, and non-hemolytic, low protein binding. Heat-sealable, bondable, sterile-weldable, and custom moldable for assemblies. This formulation is ideal for pharmaceutical, biotech, and general-purpose lab applications, including sterile filling and processing, cell harvest and media process systems, filtration, fermentation, cell and tissue culture transport. This tubing is opaque black in appearance, features a temperature range of -66 to 135°C, and offers a tubing life of approximately 1000 hours at 100 rpm.
The overall length of Viton® 97629-series stop tubing is 12" (304 mm). The legacy Ismatec® stop tubing was 180 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
£117.97 - £236.80GBP / Pkg of 12
£141.56 - £284.16GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm) Description
Availability Pricing
COMPARE
Orange/yellow 0.51 Pump Tubing, 3-Stop, FDA-Compliant, 0.51 mm ID; 12/PK
£128.22GBP / Pkg of 12
£153.86GBP (incl VAT)
COMPARE
Orange/white 0.64 Pump Tubing, 3-Stop, FDA-Compliant, 0.64 mm ID; 12/PK
£145.49GBP / Pkg of 12
£174.59GBP (incl VAT)
COMPARE
Black/Black 0.76 Pump Tubing, 3-Stop, FDA-Compliant, 0.76 mm ID; 12/PK
£147.61GBP / Pkg of 12
£177.13GBP (incl VAT)
COMPARE
Orange/orange 0.89 Pump Tubing, 3-Stop, FDA-Compliant, 0.89 mm ID; 12/PK
£151.86GBP / Pkg of 12
£182.23GBP (incl VAT)
COMPARE
White/white 1.02 Pump Tubing, 3-Stop, FDA-Compliant, 1.02 mm ID; 12/PK
£161.41GBP / Pkg of 12
£193.69GBP (incl VAT)
COMPARE
Red/red 1.14 Pump Tubing, 3-Stop, FDA-Compliant, 1.14 mm ID; 12/PK
£156.10GBP / Pkg of 12
£187.32GBP (incl VAT)
COMPARE
Gray/Gray 1.3 Pump Tubing, 3-Stop, FDA-Compliant, 1.30 mm ID; 12/PK
£157.16GBP / Pkg of 12
£188.59GBP (incl VAT)
COMPARE
Yellow/yellow 1.42 Pump Tubing, 3-Stop, FDA-Compliant, 1.42 mm ID; 12/PK
£161.41GBP / Pkg of 12
£193.69GBP (incl VAT)
COMPARE
Yellow/blue 1.52 Pump Tubing, 3-Stop, FDA-Compliant, 1.52 mm ID; 12/PK
£164.61GBP / Pkg of 12
£197.53GBP (incl VAT)
COMPARE
Blue/blue 1.65 Pump Tubing, 3-Stop, FDA-Compliant, 1.65 mm ID; 12/PK
£117.97GBP / Pkg of 12
£141.56GBP (incl VAT)
COMPARE
Green/green 1.85 Pump Tubing, 3-Stop, FDA-Compliant, 1.85 mm ID; 12/PK
£195.40GBP / Pkg of 12
£234.48GBP (incl VAT)
COMPARE
Purple/purple 2.06 Pump Tubing, 3-Stop, FDA-Compliant, 2.06 mm ID; 12/PK
£201.78GBP / Pkg of 12
£242.14GBP (incl VAT)
COMPARE
Purple/black 2.29 Pump Tubing, 3-Stop, FDA-Compliant, 2.29 mm ID; 12/PK
£198.60GBP / Pkg of 12
£238.32GBP (incl VAT)
COMPARE
Purple/orange 2.54 Pump Tubing, 3-Stop, FDA-Compliant, 2.54 mm ID; 12/PK
£227.27GBP / Pkg of 12
£272.72GBP (incl VAT)
COMPARE
Purple/white 2.79 Pump Tubing, 3-Stop, FDA-Compliant, 2.79 mm ID; 12/PK
£236.80GBP / Pkg of 12
£284.16GBP (incl VAT)
Showing 1- 15 of 15