Ismatec® Pump Tubing, 2-Stop Microbore, PharMed® BPT
Designed for optimal operation in your Ismatec pumps
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
To complete your Ismatec® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application.
The stops on the two-stop tubing are color-coded according to tubing ID. Use this tubing with any of the Ismatec® standard cartridge pumps and pump heads. Cartridges withstand pressures up to 45 psi.
The overall length of PharMed BPT 95723-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
£50.41 - £98.90GBP / Pkg of 12
£60.49 - £118.68GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm) Description
Availability Pricing
COMPARE
Orange/Red 0.19 Pump Tubing, 2-Stop, PharMed® BPT, 0.19 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Orange/Blue 0.25 Pump Tubing, 2-Stop, PharMed® BPT, 0.25 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Orange/Green 0.38 Pump Tubing, 2-Stop, PharMed® BPT, 0.38 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Orange/Yellow 0.51 Pump Tubing, 2-Stop, PharMed® BPT, 0.51 mm ID; 12/PK
£98.90GBP / Pkg of 12
£118.68GBP (incl VAT)
COMPARE
Orange/White 0.64 Pump Tubing, 2-Stop, PharMed® BPT, 0.64 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Black/Black 0.76 Pump Tubing, 2-Stop, PharMed® BPT, 0.76 mm ID; 12/PK
£89.59GBP / Pkg of 12
£107.51GBP (incl VAT)
COMPARE
Orange/Orange 0.89 Pump Tubing, 2-Stop, PharMed® BPT, 0.89 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
White/White 1.02 Pump Tubing, 2-Stop, PharMed® BPT, 1.02 mm ID; 12/PK
£50.41GBP / Pkg of 12
£60.49GBP (incl VAT)
COMPARE
Red/Red 1.14 Pump Tubing, 2-Stop, PharMed® BPT, 1.14 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Gray/Gray 1.3 Pump Tubing, 2-Stop, PharMed® BPT, 1.30 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Yellow/Yellow 1.42 Pump Tubing, 2-Stop, PharMed® BPT, 1.42 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Yellow/Blue 1.52 Pump Tubing, 2-Stop, PharMed® BPT, 1.52 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Blue/Blue 1.65 Pump Tubing, 2-Stop, PharMed® BPT, 1.65 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Green/Green 1.85 Pump Tubing, 2-Stop, PharMed® BPT, 1.85 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Purple/Purple 2.06 Pump Tubing, 2-Stop, PharMed® BPT, 2.06 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Purple/Black 2.29 Pump Tubing, 2-Stop, PharMed® BPT, 2.29 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Purple/Orange 2.54 Pump Tubing, 2-Stop, PharMed® BPT, 2.54 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
COMPARE
Purple/White 2.79 Pump Tubing, 2-Stop, PharMed® BPT, 2.79 mm ID; 12/PK
£89.41GBP / Pkg of 12
£107.29GBP (incl VAT)
Showing 1- 18 of 18