Ismatec® Microbore Pump Tubing, Solva™ Solvent/Hydrocarbon
Specifically designed for use with hydrocarbons, petroleum products and distillates
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Resists most hydrocarbons, embrittlement, and swelling
Resistant to ozone and UV light — won't crack, swell, or harden.
Translucent yellow color
Static temperature range: -37 to 74°C (-35 to 165°F)
Dynamic temperature range: 0 to 40°C (32 to 104°F)
The overall length of Solvent/Hydrocarbon extension tubing is 50 feet (15 m). The legacy Ismatec® extension tubing was 3 m long.
£50.68 - £52.49GBP / Pkg of 1
£60.82 - £62.99GBP (incl VAT)
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm)
Availability Pricing
COMPARE
0.19 1.99
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
0.25 2.05
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
0.38 2.18
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
0.51 2.31
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
0.64 2.44
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
0.76 2.46
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
0.89 2.59
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
1.02 2.72
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
1.14 2.84
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
1.3 3
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
1.42 3.12
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
1.52 3.22
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
1.65 3.35
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
COMPARE
1.85 3.55
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
2.06 3.76
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
2.29 3.99
£50.68GBP / Pkg of 1
£60.82GBP (incl VAT)
COMPARE
2.79 4.49
£52.49GBP / Pkg of 1
£62.99GBP (incl VAT)
Showing 1- 17 of 17