Ismatec® Flowmaster™ Pump
Stainless steel construction—ideal for sanitary fluid transfer
  • Housing is IP65 rated
  • Change tubing in just seconds!
  • Easy to disassemble for cleaning
This pump meets the strict hygienic requirements of the biotech, food and beverage, and pharmaceutical industries. The three-roller pump head with interchangeable rotors handles tubing with inside diameters of 6.4 to 16.0 mm and wall thicknesses of 1.6 and 3.2 mm. Accepts a 0 to 5 VDC, 0 to 10 VDC, 1 to 20 mA, or 4 to 20 mA input signal for remote speed control. Features multiple overload protection and an extensive analog interface with status information for process control systems.
£4,252.80 - £4,702.40GBP / Each
£5,103.36 - £5,642.88GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Flow Rate Per Channel (mL/min) Max Flow Rate Per Channel (mL/min) Power (VAC)
Availability Pricing
COMPARE
37 13000 115
£4,702.40GBP / Each
£5,642.88GBP (incl VAT)
COMPARE
37 13000 230
£4,252.80GBP / Each
£5,103.36GBP (incl VAT)