This product is now discontinued, View Related Products
InterCyclone Cyclonic Action Mixing System
Eliminate injuries and container damage with blade-free impeller
  • Effectively mixes high-viscosity fluids and fluids with large differences in specific gravity
  • Durable stainless steel construction resists corrosion
  • Compatible with traditional mixer motors and handheld drills
  • Small-diameter bottom creates fastest turnover, while the open bottom is more gentle
Mixer impellers require a shaft. Choose a solid or hollow core, and the length best suited to the depth of your mixing vessel.
Call For Price
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mixer Head, Anaerobic, 15 mm dia, Fully Open Bottom 316 Stainless Steel WZ-50706-00 Mfr # 15N-R-O316
COMPARE
Mixer Head, Anaerobic, 15 mm dia, Fully Open Bottom 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Anaerobic, 25 mm dia, Fully Open Bottom 316 Stainless Steel WZ-50706-01 Mfr # 25N-R-O316
COMPARE
InterCyclone Mixer Head, Anaerobic, 25 mm dia, Fully Open Bottom 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Aerobic, 25 mm dia, Fully Open Bottom 316 Stainless Steel WZ-50706-02 Mfr # 25A-R-O316
COMPARE
InterCyclone Mixer Head, Aerobic, 25 mm dia, Fully Open Bottom 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Anaerobic, 50 mm dia, Fully Open Bottom 316 Stainless Steel WZ-50706-03 Mfr # 50N-R-O316
COMPARE
InterCyclone Mixer Head, Anaerobic, 50 mm dia, Fully Open Bottom 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Anaerobic, 50 mm dia, Small dia Bottom, 316 Stainless Steel WZ-50706-04 Mfr # 50N-R-S316
COMPARE
InterCyclone Mixer Head, Anaerobic, 50 mm dia, Small dia Bottom, 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Aerobic, 50 mm dia, Fully Open Bottom, 316 Stainless Steel WZ-50706-05 Mfr # 50A-R-O316
COMPARE
InterCyclone Mixer Head, Aerobic, 50 mm dia, Fully Open Bottom, 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Aerobic, 50 mm dia, Small dia Bottom, 316 Stainless Steel WZ-50706-06 Mfr # 50A-R-S316
COMPARE
InterCyclone Mixer Head, Aerobic, 50 mm dia, Small dia Bottom, 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Anaerobic 100 mm dia, Fully Open Bottom 316 Stainless Steel WZ-50706-07 Mfr # 101N-R-O316
COMPARE
InterCyclone Mixer Head, Anaerobic 100 mm dia, Fully Open Bottom 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Anaerobic, 100 mm dia, Small dia Bottom 316 Stainless Steel WZ-50706-08 Mfr # 101N-R-S316
COMPARE
InterCyclone Mixer Head, Anaerobic, 100 mm dia, Small dia Bottom 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Aerobic, 100 mm dia, Fully Open Bottom, 316 Stainless Steel WZ-50706-09 Mfr # 101A-R-O316
COMPARE
InterCyclone Mixer Head, Aerobic, 100 mm dia, Fully Open Bottom, 316 Stainless Steel
Call For Price
InterCyclone Mixer Head, Aerobic, 100 mm dia, Small dia Bottom, 316 Stainless Steel WZ-50706-10 Mfr # 101A-R-S316
COMPARE
InterCyclone Mixer Head, Aerobic, 100 mm dia, Small dia Bottom, 316 Stainless Steel
Call For Price
Showing 1- 11 of 11