Inlet Filters/Silencers
Protects oilless air compressors and regenerative blowers up to 99%+ efficiency
  • Interchangeable polyester and paper filter elements to suit various applications
  • Durable seamless housings — no welds to rust or vibrate apart
  • Filters available with silencing tubes to minimize noise levels and pressure drop
Inlet filter assemblies remove impurities in the airline before entering your air compressor or regenerative blower. Paper filter elements provide a 2 micron level efficiency while polyester filter elements provide an efficiency level down to 5 microns. Inlet filter/silencer assemblies are able to mount either vertically or horizontally.
£34.24GBP / Each
£41.09GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Port Size
Availability Pricing
COMPARE
12 1/2\" NPT(M)
£34.24GBP / Each
£41.09GBP (incl VAT)