North Slope Chillers Portable Industrial Chillers
Maintains safe temperatures for critical materials and equipment
  • Simple to operate, install, and relocate
  • Four mounted casters allow for easy mobility (not available on Frost models)
  • Brazed plate heat exchanger
  • Designed for indoor operation
  • Air-cooled, closed-loop fluid chillers
These portable air-cooled stand-alone industrial chillers, equipped with a continuous duty, non-ferrous piping system, easily remove heat from your materials and transfer it to the surrounding ambient air. Designed for indoor operation, the chillers feature a hermetically sealed compressor, brazed plate heat exchanger, fluid level gauge, pressure gauge, digital controller, and powder-coated steel cabinet.
£3,713.96 - £9,633.48GBP / Each
£4,456.75 - £11,560.18GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (L/min) Min Temperature (° C) Description
Availability Pricing
North Slope Chillers Industrial Portable Chiller, 1/3 Ton, 4,000 BTU's/hr Capacity WZ-42024-95 Mfr # NSC0330-110/1
COMPARE
22.7 -9.5 Industrial Portable Chiller, 1/3 Ton, 4,000 BTU's/hr Capacity
£3,713.96GBP / Each
£4,456.75GBP (incl VAT)
North Slope Chillers Industrial Portable Chiller, 1/2 Ton, 6,000 BTU's/hr Capacity WZ-42025-20 Mfr # NSC0500110/1
COMPARE
15 1.7 Industrial Portable Chiller, 1/2 Ton, 6,000 BTU's/hr Capacity
£5,517.74GBP / Each
£6,621.29GBP (incl VAT)
North Slope Chillers Industrial Portable Chiller, 1 Ton, 12,000 BTU's/hr Capacity WZ-42025-21 Mfr # NSC1000230/1
COMPARE
15 4.4 Industrial PortableChiller, 1 Ton, 12,000 BTU's/hr Capacity
£7,382.81GBP / Each
£8,859.37GBP (incl VAT)
North Slope Chillers Industrial Portable Chiller, 1 Ton, 11,900 BTU's/hr Capacity WZ-42025-22 Mfr # NSC1000LT230/1
COMPARE
15 -12.2 Industrial Portable Chiller, 1 Ton, 11,900 BTU's/hr Capacity
£7,751.39GBP / Each
£9,301.67GBP (incl VAT)
North Slope Chillers Industrial Portable Chiller, 2 Ton, 25,400 BTU's/hr Capacity WZ-42025-23 Mfr # NSC2000230/1
COMPARE
170 4.4 Industrial Portable Chiller, 2 Ton, 25,400 BTU's/hr Capacity
£8,852.90GBP / Each
£10,623.48GBP (incl VAT)
North Slope Chillers Industrial Portable Chiller, 2 Ton, 27,200 BTU's/hr Capacity WZ-42025-24 Mfr # NSC2000LT230/1
COMPARE
170 -12.2 Industrial Portable Chiller, 2 Ton, 27,200 BTU's/hr Capacity
£9,633.48GBP / Each
£11,560.18GBP (incl VAT)