This product is now discontinued, View Related Products

iMSO-104 Oscilloscope Accessories

Choose from our comprehensive line of Oscium iMSO-104 Oscilloscope Accessories and other Accessories for Oscilloscopes products.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Bandwidth (MHz) Channels Input Impedance
Availability Pricing
Oscium OSC3056 Logic Harness, , 9 Pin with SMD Grabbers WZ-20044-19 Mfr # OSC3056
COMPARE
Call For Price
Oscium OSC1100 Analog Probe, 1x / 10x WZ-20044-22 Mfr # OSC1100
COMPARE
Call For Price
Oscium OSC3055 Logic Harness, 5 Pin with SMD Grabbers WZ-20044-23 Mfr # OSC3055
COMPARE
Call For Price
Oscium OSC3050 Logic Harness Oscilloscope Accessory, 5 Pin WZ-20044-24 Mfr # OSC3050
COMPARE
Call For Price
Oscium OSC3005 Oscilloscope SMD Grabbers WZ-20044-25 Mfr # OSC3005
COMPARE
Call For Price
Oscium OSC1109 Extension cable, 19 in. WZ-20044-26 Mfr # OSC1109
COMPARE
Call For Price
Oscium OSC9001 Oscilloscope Screwdriver WZ-20044-27 Mfr # OSC9001
COMPARE
Call For Price
Oscium OSC1101 Conducted Measurement Kit, for WiPry-Combo and WiPry-Power Modules WZ-20044-29 Mfr # OSC1101
COMPARE
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat