This product is now discontinued, View Related Products
Imada High Performance Digital Force Gauges With Remote Sensor
Remote sensor is ideal for use in confined spaces or hard to reach applications
  • RS-232, Digimatic, and analog outputs
  • ±0.2% full scale, ±1 digit
  • Process 1000 data/second – not just the peak
  • Programmable high/low setpoints with color coded LEDs and output signal for go/no go testing
  • Reversible display
  • Overload capacity of 200%
  • Real time and peak mode (selectable)
  • Overload alarm and output signal
  • Runs on internal NiMH batteries or AC adapter
Call For Price
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Imada Z2S-DPU-0.4 Digital Force Gauge with Remote Sensor, 7 ozf. WZ-25301-35 Mfr # ZTS-DPU-0.4
COMPARE
Digital Force Gauge with Remote Sensor, 7 ozf.
Call For Price
Imada Z2S-DPU-1 Digital Force Gauge with Remote Sensor, 18.00 ozf. WZ-25301-36 Mfr # ZTS-DPU-1
COMPARE
Digital Force Gauge with Remote Sensor, 18.00 ozf.
Call For Price
Imada Z2S-DPU-4 Digital Force Gauge with Remote Sensor, 4.400 lbf. WZ-25301-37 Mfr # ZTS-DPU-4
COMPARE
Digital Force Gauge with Remote Sensor, 4.400 lbf.
Call For Price
Imada Z2S-DPU-11 Digital Force Gauge with Remote Sensor, 11.00 lbf. WZ-25301-38 Mfr # ZTS-DPU-11
COMPARE
Digital Force Gauge with Remote Sensor, 11.00 lbf.
Call For Price
Imada Z2S-DPU-44 Digital Force Gauge with Remote Sensor, 44.00 lbf. WZ-25301-39 Mfr # ZTS-DPU-44
COMPARE
Digital Force Gauge with Remote Sensor, 44.00 lbf.
Call For Price
Imada Z2S-DPU-110 Digital Force Gauge with Remote Sensor, 110.0 lbf. WZ-25301-40 Mfr # ZTS-DPU-110
COMPARE
Digital Force Gauge with Remote Sensor, 110.0 lbf.
Call For Price
Imada Z2S-DPU-220 Digital Force Gauge with Remote Sensor, 220.0 lbf. WZ-25301-41 Mfr # ZTS-DPU-220
COMPARE
Digital Force Gauge with Remote Sensor, 220.0 lbf.
Call For Price
Imada Z2S-DPU-440 Digital Force Gauge with Remote Sensor, 440.0 lbf. WZ-25301-42 Mfr # Z2S-DPU-440
COMPARE
Digital Force Gauge with Remote Sensor, 440.0 lbf.
Call For Price
Imada Z2S-DPU-1100 Digital Force Gauge with Remote Sensor, 1100 lbf. WZ-25301-43 Mfr # ZTS-DPU-1100
COMPARE
Digital Force Gauge with Remote Sensor, 1100 lbf.
Call For Price
Imada Z2S-DPU-2200 Digital Force Gauge with Remote Sensor, 2200 lbf. WZ-25301-44 Mfr # ZTS-DPU-2200
COMPARE
Digital Force Gauge with Remote Sensor, 2200 lbf.
Call For Price
Imada Z2S-DPU-4400 Digital Force Gauge with Remote Sensor, 4400 lbf. WZ-25301-45 Mfr # ZTS-DPU-4400
COMPARE
Digital Force Gauge with Remote Sensor, 4400 lbf.
Call For Price
Showing 1- 11 of 11