IKA® T25 easy clean digital Homogenizer Rotor-Stator Generators
Quick and easy cleaning of the rotor-stator generators—no disassembling required
  • Rotor-stator generators cleaning status allows for quick cleaning saving on down time
  • Choose from systems with or without temperature measurement
The T25 easy clean system is ideal for batch homogenizing of cell tissues, emulsifying suspended solids, and air pollution filter extraction. The compatible rotor-stator generators feature cleaning status indicator to inform you of when cleaning is required. Cleaning is quick and easy—no disassembling required. Shielded in a 316 stainless steel shaft, the rotor acts as a centrifugal pump to recirculate the liquid and suspended solids through the generator, where shear, impact, collision, and cavitation provide rapid homogenization.
The T25 Control homogenizer along with the 04739-25 rotor stator generator allows continuous temperature measurement with an integrated temperature sensor. Device overheating is prevented by continuous temperature monitoring and the setting of temperature limits.
Order homogenizers separately.
£2,332.40GBP / Each
£2,798.88GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Min Sample Size (mL) Description
Availability Pricing
IKA S 25 Homogenizer Rotor-Stator Generator with Integrated Temperature Sensor, 50-150 mm depth, 10-1500 mL WZ-04739-25 Mfr # 0025007398
COMPARE
1500 10 Homogenizer Rotor-Stator Generator with Integrated Temperature Sensor, 50-150 mm depth, 10-1500 mL
£2,332.40GBP / Each
£2,798.88GBP (incl VAT)