This product is now discontinued, View Related Products
IKA RV 8 Rotary Evaporators
Economical option for standard evaporating applications
  • Easy-to-use manual lift with integrated safety lift-out function
  • Ambidextrous design makes operation more comfortable
  • Adjustable immersion angle and height for maximum heating efficiency
  • Compatible with complete range of IKA rotary evaporator glassware
The RV 8 vertical distilling system is designed for ease of use in all standard evaporating applications. The manual lift with ambidextrous design lets you precisely position the glassware. Digital displays indicate both rotation speed and heating bath temperature, allowing for optimal control of the process. Easy-to-use clamp system simplifies changing evaporator flasks.
Ergonomic 3 L water/oil heating bath has a locking function for temperature settings from ambient to 180°C. Bath is designed with integrated ergonomic carrying handles for convenient positioning and oblong design for fast heating.
Systems are available with standard glassware, with dry-ice condenser glassware, or with no glassware. Safety-coated glassware options are available; they are resistant to acids and solvents and protect against shattered glass.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Condenser Type Glass Type Description
Availability Pricing
IKA RV 8 V Rotary Evaporator with Uncoated Glassware WZ-28711-03 Mfr # 10003570
COMPARE
Vertical Uncoated Rotary Evaporator with Uncoated Glassware
Call For Price
IKA RV 8 V-C Rotary Evaporator with Safety-Coated Glassware WZ-28711-04 Mfr # 10003571
COMPARE
Vertical Safety-coated Rotary Evaporator with Safety-Coated Glassware
Call For Price
IKA RV 8 FLEX Rotary Evaporator, Vertical, No Glassware WZ-28711-05 Mfr # 10003572
COMPARE
None Uncoated Rotary Evaporator, Vertical, No Glassware
Call For Price
IKA RV 8 Rotary Evaporator with Dry Ice Condenser, Uncoated Glassware WZ-28711-06 Mfr # 10003573
COMPARE
Cold-trap Uncoated Rotary Evaporator with Dry Ice Condenser, Uncoated Glassware
Call For Price
IKA RV 8 Rotary Evaporator with Dry Ice Condenser, Saftey-Coated Glassware WZ-28711-07 Mfr # 10003574
COMPARE
Cold-trap Safety-coated Rotary Evaporator with Dry Ice Condenser, Saftey-Coated Glassware