This product is now discontinued, View Related Products
IKA® RV 3 Basic Rotary Evaporators
Safe, innovative design features digital displays for speed and temperature
  • Integrated safety function lifts flask out of bath in case of power failure
  • Red warning light indicates unlocked position of vapor tube
  • Single-handed manual lift offers precise positioning of glassware
  • Low device voltage (24 V) ensures user safety
  • Adjustable immersion angle
These rotary evaporators are designed and engineered for ease of use, while maintaining the highest level of safety. Manual lift handling can be done with one hand for precise positioning of glassware and is suitable for left- or right-handed operators. A flask clamping mechanism with integrated push-off function allows for easy exchange of evaporation flasks. The transparent vertical condenser's small footprint makes efficient use of the 1500 cm² cooling surface.
The unique 3 L bath design provides fast heat up, allowing for quicker experiments. Non-exposed heating coils and integrated handles offer safe operation. Bath heats from 20 to 90°C, adding versatility by allowing use of water or oil. The DC motor provides rotation speed from 20 to 300 rpm. Each unit comes standard with a dual digital display for both bath temperature and rotational speed to provide more control and easier repetition of experiments.
The evaporators are available with or without safety-coated glassware to minimize breakage and increase safety. Safety coating resists most solvents and acids while protecting you against shattered glass and dangerous reagents due to broken glass.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Bath Type Condenser Type Glass Type
Availability Pricing
IKA RV 3 V Dual Rotary Evaporator With Vertical Uncoated Glassware, 100 to 240 VAC WZ-28710-81 Mfr # 10003491
COMPARE
Water/Oil Vertical Uncoated
Call For Price
IKA RV 3 V-C Dual Rotary Evaporator With Vertical Safety Coated Glassware, 100 to 240 VAC WZ-28710-83 Mfr # 10003492
COMPARE
Water Vertical Safety-coated
Call For Price
IKA RV 3 Dual Rotary Evaporator With Dry Ice Condenser Uncoated Glassware, 100 to 240 VAC WZ-28710-85 Mfr # 10003494
COMPARE
Water/Oil Cold-trap Uncoated
Call For Price
IKA RV 3 Dual Rotary Evaporator With Dry Ice Condenser Safety Coated Glassware, 100 to 240 VAC WZ-28710-86 Mfr # 10003495
COMPARE
Water/Oil Cold-trap Safety-coated
Call For Price
IKA RV 3 FLEX Dual Entry-Level Rotary Evaporator, 100 to 240 VAC WZ-28710-87 Mfr # 10003493
COMPARE
Water Vertical Uncoated