This product is now discontinued, View Related Products
IEQ Chek Indoor Air Quality Monitors
Large, multiline LCD shows all installed sensor readings simultaneously
  • Internal draw pump facilitates sampling
These handheld monitors measure temperature, humidity, and up to five additional plug-and-play smart sensors—ideal for indoor environmental quality measurements. Display all installed sensors simultaneously on the large, backlit display. Operate as a handheld unit, stand upright on a flat surface, fasten to a wall, or screw a tripod onto the bottom thread. Program alarm set points for any installed sensor; an audible alarm alerts you when the set point has been reached.
Call For Price
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Max CO2 (ppm) Max CO2 (%) Description
Availability Pricing
Bacharach IEQ Chek Indoor Air Quality Monitor, CO2; Pump WZ-81973-41 Mfr # 1510-0002
COMPARE
5000 Indoor Air Quality Monitor, CO2; Pump
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO2; Pump/Data logger WZ-81973-42 Mfr # 1510-0004
COMPARE
5000 Indoor Air Quality Monitor, CO2; Pump/Data logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor with CO2 and CO sensors WZ-81973-43 Mfr # 1510-1001
COMPARE
5000 Indoor Air Quality Monitor with CO2 and CO sensors
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO; Logger WZ-81973-44 Mfr # 1510-1003
COMPARE
5000 Indoor Air Quality Monitor, CO; Logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO/CO2; Pump WZ-81973-45 Mfr # 1510-1002
COMPARE
5000 Indoor Air Quality Monitor, CO/CO2; Pump
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO/CO2; Pump/Data logger WZ-81973-46 Mfr # 1510-1004
COMPARE
5000 Indoor Air Quality Monitor, CO/CO2; Pump/Data logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO/CO2/NO2; Pump WZ-81973-47 Mfr # 1510-1302
COMPARE
5000 Indoor Air Quality Monitor, CO/CO2/NO2; Pump
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO/CO2/NO2; Pump/Data logger WZ-81973-48 Mfr # 1510-1304
COMPARE
5000 Indoor Air Quality Monitor, CO/CO2/NO2; Pump/Data logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO/CO2/CH2O; Logger WZ-81973-49 Mfr # 1510-1403
COMPARE
5000 Indoor Air Quality Monitor, CO/CO2/CH2O; Logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO/CO2/TVOC; Pump/Logger WZ-81973-51 Mfr # 1512-1008
COMPARE
5000 Indoor Air Quality Monitor, CO/CO2/TVOC; Pump/Logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, High-Range CO2; Pump WZ-81973-52 Mfr # 1520-0002
COMPARE
10000 Indoor Air Quality Monitor, High-Range CO2; Pump
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, High-Range CO2; Logger WZ-81973-53 Mfr # 1520-0003
COMPARE
10000 Indoor Air Quality Monitor, High-Range CO2; Logger
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, 0-5% CO2 WZ-81973-54 Mfr # 1530-0001
COMPARE
5 Indoor Air Quality Monitor, 0-5% CO2
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, 0-20% CO2; Pump WZ-81973-55 Mfr # 1540-0002
COMPARE
20 Indoor Air Quality Monitor, 0-20% CO2; Pump
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO2; Pump/Remote WZ-81973-56 Mfr # 1540-0009
COMPARE
20 Incubator Air Quality Monitor, CO2; Pump/Remote
Call For Price
Bacharach IEQ Chek Indoor Air Quality Monitor, CO2/O2; Pump/Remote WZ-81973-57 Mfr # 1540-2009
COMPARE
20 Incubator Air Quality Monitor, CO2/O2; Pump/Remote
Call For Price
Showing 1- 16 of 16