I/P Standard Pump Heads
Best accuracy and repeatability of any I/P pump head—ideal for dispensing and metering
  • Each model is optimized for a single tube size to maximize performance and tubing life
  • Cost effective pump head is an excellent choice for dedicated applications
  • Stack two pump heads to double flow rate
The Standard pump head’s classic design ensures the most accurate and repeatable performance—important in dispensing applications. A 360-degree precision molded occlusion body ensures optimum accuracy and long tubing life.
Select from PC housing with CRS or SS rotor, or PPS housing with SS rotor. PC is economical and transparent, while PPS offers enhanced chemical resistance. SS rotors feature PTFE-sealed SS bearings for long life and reduced maintenance; CRS rotors have shielded bearings.
£461.58 - £816.36GBP / Each
£553.90 - £979.63GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Housing Material Rotor Material Tubing sizes accepted
Availability Pricing
COMPARE
Polycarbonate (PC) Cold-rolled Steel I/P® 73
£461.58GBP / Each
£553.90GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel I/P® 73
£642.59GBP / Each
£771.11GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel I/P® 82
£689.66GBP / Each
£827.59GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel I/P® 82
£499.59GBP / Each
£599.51GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel I/P® 73
£769.30GBP / Each
£923.16GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel I/P® 82
£816.36GBP / Each
£979.63GBP (incl VAT)