This product is now discontinued, View Related Products
Hydraulic Press Test Systems, Automatic
Test the physical limits of your sample – systems are ideal for a wide range of applications including material research and quality control
  • Compact design efficiently uses your laboratory bench space
  • Cast-iron base and bolsters with two threaded columns provide easy daylight adjustment
These heavy-duty systems handle a wide range of laboratory quality and performance applications such as compression, flexural, or shear strength; flow and color dispersion; and material research using ceramics, composites, construction materials, resins, and more. Safety features include a 10-gauge steel shield with see-through, shatterproof polycarbonate access door. The base has holes for permanent bench mounting and for optional permanent bench mounting.
Automatic Hydraulic Presses feature a touch-screen interface allowing you to select auto or manual mode, and units in lb, US tons, metric tons, or kg. Scroll through menu to enter parameters for pressure, pump speed, and dwell time. Presses feature audible/visual alarms and proximity switch for adjustable slowdown position. Requires two-hand operation for maximum safety.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Platen Included Power (VAC)
Availability Pricing
Carver 3888.1NE0 Hydraulic Press Test System, Automatic, 115 VAC, 60 Hz WZ-59620-30 Mfr # 3888.1PL0
COMPARE
None 115
Call For Price
Carver 3888.4NE0 Hydraulic Press Test System, Automatic, 230 VAC, 60 Hz WZ-59620-35 Mfr # 3888.4PL0
COMPARE
None 230
Call For Price
Carver 3889.1NE0 Hydraulic Press Test System, Automatic with Heated Platen, 115 VAC, 60 Hz WZ-59620-40 Mfr # 3889.1PL0
COMPARE
Yes, two heated 115