This product is now discontinued, View Related Products

Huber Grande Fleur Heating/Cooling Recirculators

Precise and reproducible temperature control results for closed-loop applications
  • Environmentally friendly temperature control units use natural refrigerants
  • Electronic, user-adjustable overtemperature protection and optical low-level sensor
  • Intuitive Pilot ONE touch-screen controller simplifies tasks
The Grand Fleur series of circulators offers the widest temperature range without requiring a fluid change, as well as fast heating and cooling rates. These circulators use environmentally friendly natural refrigerants, air or water depending on the model, while boasting a powerful, variable-speed pressure pump—ideal for the control of research reactors.

The integrated Pilot ONE controller features a large, color touch-screen display with multilanguage support, convenient color-coded navigation system, and USB and Ethernet ports for communication and remote control through your tablet, PC, or network. Additional controller capabilities provide further functionality and include programmability (10 programs, 100 steps each), ramping (linear and nonlinear), five-point calibration, and the option to control the system based on the internal or process temperature.
Call For Price

11 variations of this product are available.

See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
Huber 1041.0004.01 Heating/Cooling Recirculator, Air-Cooled, Open; 230VAC/50Hz WZ-12125-77 Mfr # 1041.0004.01
COMPARE
230 Heating/Cooling Recirculator, Air-Cooled, Open; 230VAC/50Hz
Call For Price
Huber Grande Fleur Heating/Cooling Recirculator, Air-Cooled, Sealed; 115VAC/60Hz WZ-12125-90 Mfr # 1041.0002.01
COMPARE
115 Heating/Cooling Recirculator, Air-Cooled, Sealed; 115VAC/60Hz
Call For Price
Huber 1041.0011.01 Heating/Cooling Recirculator, Water-Cooled, Open; 115VAC/50Hz WZ-12125-91 Mfr # 1041.0011.01
COMPARE
115 Heating/Cooling Recirculator, Water-Cooled, Open; 115VAC/50Hz
Call For Price
Huber 1041.0012.01 Heating/Cooling Recirculator, Water-Cooled, Open; 208VAC/60Hz WZ-12125-92 Mfr # 1041.0012.01
COMPARE
208 Heating/Cooling Recirculator, Water-Cooled, Open; 208VAC/60Hz
Call For Price
Huber 1041.0005.01 Heating/Cooling Recirculator, Air-Cooled, Open; 115VAC/60Hz WZ-12125-93 Mfr # 1041.0005.01
COMPARE
115 Heating/Cooling Recirculator, Air-Cooled, Open; 115VAC/60Hz
Call For Price
Huber 1041.0006.01 Heating/Cooling Recirculator, Air-Cooled, Open; 208VAC/60Hz WZ-12125-94 Mfr # 1041.0006.01
COMPARE
208 Heating/Cooling Recirculator, Air-Cooled, Open; 208VAC/60Hz
Call For Price
Huber Grande Fleur W Heating/Cooling Recirculator, Water-Cooled, Sealed; 115VAC/60Hz WZ-12125-95 Mfr # 1041.0008.01
COMPARE
115 Heating/Cooling Recirculator, Water-Cooled, Sealed; 115VAC/60Hz
Call For Price
Huber Grande Fleur W Heating/Cooling Recirculator, Water-Cooled, Sealed; 208VAC/60Hz WZ-12125-96 Mfr # 1041.0009.01
COMPARE
208 Heating/Cooling Recirculator, Water-Cooled, Sealed; 208VAC/60Hz
Call For Price
Huber Grande Fleur Heating/Cooling Recirculator, Air-Cooled, Sealed; 230VAC/50Hz WZ-12125-97 Mfr # 1041.0001.01
COMPARE
230 Heating/Cooling Recirculator, Air-Cooled, Sealed; 230VAC/50Hz
Call For Price
Huber 1041.0010.01 Heating/Cooling Recirculator, Water-Cooled, Open; 230VAC/50Hz WZ-12125-98 Mfr # 1041.0010.01
COMPARE
230 Heating/Cooling Recirculator, Water-Cooled, Open; 230VAC/50Hz
Call For Price
Huber Grande Fleur W Heating/Cooling Recirculator, Water-Cooled, Sealed; 230VAC/50Hz WZ-12125-99 Mfr # 1041.0007.01
COMPARE
230 Heating/Cooling Recirculator, Water-Cooled, Sealed; 230VAC/50Hz
Call For Price
Showing 1- 11 of 11

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat