This product is now discontinued, View Related Products

Huber CC Series Refrigerated Heating Circulator Baths

No more hard-to-follow instructions—icon-driven menu is intuitive and easy!
  • Color-coded menu categories simplify workflow
  • Favorites menu provides quick access to frequently used options
  • Software wizards guide you through setup to ensure correct settings
  • Large 5.7" (14.5 cm) color TFT touchscreen display is user friendly
  • USB port allows connection with devices
These cooling heating circulators feature a modern controller with advanced functions for trouble-free setup. All important information and program parameters are accessible with a few keystrokes saving you valuable time. The large touchscreen display, color-coded menu, and setup wizard ensure fool-proof programming. Ethernet, USB, and RS-232 interfaces provide network, device, and PC connectivity for remote setup and monitoring.

Air- and water-cooled options, bath sizes from benchtop to floor standing, and a wide array of heating wattages and cooling capacities are available.
Call For Price

29 variations of this product are available.

See All Product Options Below
29 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Huber CC-K6 Refrigerated Heating Bath, 110 VAC, 50/60Hz WZ-12751-67 Mfr # 2008.0006.01
COMPARE
200 110 4.5
Call For Price
Huber CC-K6s Refrigerated Heating Bath, 110 VAC, 50/60Hz WZ-12751-68 Mfr # 2008.0004.01
COMPARE
260 110 4.5
Call For Price
Huber CC-K12 Refrigerated Heating Bath, 115 VAC, 60Hz WZ-12751-69 Mfr # 2009.0004.01
COMPARE
250 115 12
Call For Price
Huber CC-K15 Refrigerated Heating Bath, 115 VAC, 60Hz WZ-12751-70 Mfr # 2010.0004.01
COMPARE
250 115 15
Call For Price
Huber CC-K20 Refrigerated Heating Bath, 115 VAC, 60Hz WZ-12751-71 Mfr # 2011.0004.01
COMPARE
400 115 20
Call For Price
Huber CC-K25 Refrigerated Heating Bath, 115 VAC, 60Hz WZ-12751-72 Mfr # 2012.0004.01
COMPARE
400 115 25
Call For Price
Huber CC-405 Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-73 Mfr # 2017.0003.01
COMPARE
700 208 5
Call For Price
Huber CC-405w Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-74 Mfr # 2017.0004.01
COMPARE
700 208 5
Call For Price
Huber CC-410 Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-75 Mfr # 2019.0006.01
COMPARE
800 208 22
Call For Price
Huber CC-410wl Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-76 Mfr # 2019.0002.01
COMPARE
800 208 22
Call For Price
Huber CC-415 Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-77 Mfr # 2018.0007.01
COMPARE
1200 208 5
Call For Price
Huber CC-415wl Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-78 Mfr # 2018.0009.01
COMPARE
1200 208 5
Call For Price
Huber CC-505 Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-79 Mfr # 2018.0010.01
COMPARE
1200 208 5
Call For Price
Huber CC-505wl Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-80 Mfr # 2018.0011.01
COMPARE
1200 208 5
Call For Price
Huber CC-508 Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-81 Mfr # 2018.0014.01
COMPARE
1500 208 5
Call For Price
Huber CC-508w Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-82 Mfr # 2018.0015.01
COMPARE
1500 208 5
Call For Price
Huber CC-510w Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-83 Mfr # 2020.0005.01
COMPARE
2100 208 18
Call For Price
Huber CC-515 Refrigerated Heating Circulator Bath, 460 VAC, 60Hz WZ-12751-84 Mfr # 2021.0002.01
COMPARE
3300 460 26
Call For Price
Huber CC-515 Refrigerated Heating Circulator Bath, 208 VAC, 60Hz WZ-12751-85 Mfr # 2021.0003.01
COMPARE
3300 208 26
Call For Price
Huber CC-515w Refrigerated Heating Circulator Bath, 460 VAC, 60Hz WZ-12751-86 Mfr # 2020.0006.01
COMPARE
3300 460 18
Call For Price
Showing 1 - 20 of 29

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat