Kimble® ULTRA-WARE® HPLC Mobile Phase Reservoirs
Chemically inert glass prevents leaching of any extractables into the mobile phase solvents
  • Constructed of durable safety coated glass
These reservoirs are manufactured of high-quality borosilicate glass for maximum chemical resistance. Reservoirs are coated with plastic for safety and UV resistance. Select from conical bottom configuration for maximum solvent utilization or flat bottom reservoirs.
£297.09 - £814.64GBP / Each
£356.51 - £977.57GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bottom Type
Availability Pricing
DWK Life Sciences (Kimble) HPLC Mobile Phase Reservoir, Glass, Conical Bottom; 5.0 Liter WZ-02017-15 Mfr # 953901-5002
COMPARE
5000 Conical
£814.64GBP / Each
£977.57GBP (incl VAT)
DWK Life Sciences (Kimble) HPLC Mobile Phase Reservoir, Glass, Flat Bottom; 2.0 Liter WZ-02017-25 Mfr # 953902-2002
COMPARE
2000 Flat
£297.09GBP / Each
£356.51GBP (incl VAT)