Hirschmann Solarus Electronic Burettes (Burets)
Operates on solar power – no more charging or changing batteries
  • High accuracy
  • Burette (buret) head rotates 360° for better viewing
  • Manipulate and report data easily with RS-232 output
These digital burettes (burets) are designed for titrating in water treatment plants, material testing facilities and analytical laboratories. Integral solar panels eliminate the need for charging or changing batteries and allow for enhanced portability even in limited-lighting conditions. Easy-to-use recirculating system quickly primes the burette reducing loss of valuable reagents. Features a large LCD, user-friendly keypad, Quick-Cal individual calibration and indicator arrows to provide filling and titration direction. The hand wheels are equipped with a soft touch surface and smooth opertaing gear ensuring precise delivery of your solution. The RS-232 interface allows you to download data to your computer. RS-232 port included for computer interface. Interface cable sold separately. Valves are easily dismantled and exchangeable with provided wrench. Note: Not for use with hydrofluoric acid. Autoclavable to 250°F (121°C). What's included:recirculating tube, suction tube, thread adapters, and individual test certificate
£1,391.51GBP / Each
£1,669.81GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance Description
Availability Pricing
Hirschmann 9392020 Solarus Electronic Burette, Bottletop Dispenser; 20 mL WZ-25503-20 Mfr # 9392020
COMPARE
20 0.2 Solarus Electronic Burette, Bottletop Dispenser; 20 mL
£1,391.51GBP / Each
£1,669.81GBP (incl VAT)
Hirschmann 9392050 Solarus Electronic Burette, Bottletop Dispenser; 50 mL WZ-25503-25 Mfr # 9392050
COMPARE
50 0.2 Solarus Electronic Burette, Bottletop Dispenser; 50 mL
£1,391.51GBP / Each
£1,669.81GBP (incl VAT)