High-Tech Conversions Eco-Friendly Green Critical Task Wipes
Eco-friendly lab wipes made from 100% recycled cellulose
  • Ideal for optics and other sensitive surfaces
  • Perfect wipe for large surface areas
Made from virgin cellulose, these lab wipes are excellent for your light-duty applications, will not scratch, and can be used in a variety of industries. The wipes are made of single-ply, very absorbent, and soft material making them ideal for cleaning optics and sensitive instruments and equipment in your lab facility. Since they are low linting and low in chemical extractables, they’ll leave surfaces extremely clean.
The 15? x 16.6? (38 x42 cm) wipes are packaged 140 per box.
£13.80GBP / Pkg of 140
£16.56GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
High-Tech Conversions Critical Task Wipe, 15 x 16.6? Single-Ply Cellulose; 140 Wipes/Box WZ-78902-96 Mfr # Techwipe-XL
COMPARE
16 19/32 15 Friendly Green Critical Task Wipe, 15″ x 16.6″ Single-Ply Cellulose; 140 Wipes/Box
£13.80GBP / Pkg of 140
£16.56GBP (incl VAT)
High-Tech Conversions Friendly Green Critical Task Wipe, 15″ x 16.6″ Single-Ply Cellulose; 140 Wipes/Box, 15 Boxes/CS WZ-78902-98 Mfr # FG-2
COMPARE
16 19/32 15 Friendly Green Critical Task Wipe, 15″ x 16.6″ Single-Ply Cellulose; 140 Wipes/Box, 15 Boxes/CS