High-Purity Two-Stage Gas Regulators
Ideal for corrosive or noncorrosive gas applications. Designed and constructed for diffusion resistance and easy cleanup. Only brass, stainless steel (SS), PTFE, and Kel-F® contact the operating media. Metal diaphragm, packless valves, and high-purity seals minimize or eliminate outgoing and inboard diffusion. Supplied with SS screen filter (25 µm); and brass body, bonnet, and gauges. Two-Stage High-Purity Regulators let you precisely regulate working pressure.
£725.91 - £864.20GBP / Each
£871.09 - £1,037.04GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (SCFH) Control Range (high) (PSI) Max Inlet Pressure (PSI)
Availability Pricing
COMPARE
450 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
450 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
450 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
450 3000
£815.40GBP / Each
£978.48GBP (incl VAT)
COMPARE
450 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
900 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
900 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
900 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
COMPARE
900 3000
£864.20GBP / Each
£1,037.04GBP (incl VAT)
COMPARE
900 3000
£725.91GBP / Each
£871.09GBP (incl VAT)
Showing 1- 10 of 10