High-Flow Quick Disconnect Couplings (HFC series)
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect—fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Designed to enable enhanced flow in a coupling with a 3/8" flow path, the HFC series is available in polypropylene and polysulfone for a wide variety of applications. The HFC series has an ergonomic, easy-to-operate design and comes standard with a shrouded latch to protect against accidental disconnect. UV-resistant polysulfone Couplings of UV-resistant polysulfone and with BSP threaded terminations are also available; contact Cole-Parmer for more information.
£7.34 - £186.90GBP / Each
£8.81 - £224.28GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Description
Availability Pricing
CPC (Colder) HFC12612 Coupling Body, Panel Mount, Polypropylene (PP), 3/8" OD Tube WZ-31303-77 Mfr # HFC12612
COMPARE
Polypropylene Quick-Disconnect x Compression Coupling Body, Panel Mount, 3/8" OD Tube
£14.46GBP / Each
£17.35GBP (incl VAT)
CPC (Colder) HFCD20812 High-Flow Quick-Disconnect, Valved Polypropylene (PP) Insert, 1/2" OD WZ-31303-86 Mfr # HFCD20812
COMPARE
Polypropylene Quick-Disconnect x Compression High-Flow Quick-Disconnect, Valved PP Insert, 1/2" OD
£11.60GBP / Each
£13.92GBP (incl VAT)
CPC (Colder) HFCD101235GHT High-Flow Quick-Disconnect, Valved Polysulfone (PS) Body, 3/4" GHT(M) WZ-31307-57 Mfr # HFCD101235GHT
COMPARE
Polysulfone Quick-Disconnect x GHT High-Flow Quick-Disconnect, Valved PS Body, 3/4" GHT(M)
£14.27GBP / Each
£17.12GBP (incl VAT)
CPC (Colder) HFCD241235GHT High-Flow Quick-Disconnect, Valved Polysulfone (PS) Insert, 3/4" GHT(M) WZ-31307-59 Mfr # HFCD241235GHT
COMPARE
Polysulfone Quick-Disconnect x GHT High-Flow Quick-Disconnect, Valved PS Insert, 3/4" GHT(M)
£11.98GBP / Each
£14.38GBP (incl VAT)
CPC (Colder) HFC241235GHT High-Flow Quick-Disconnect, Non-Valved Polysulfone (PS) Insert, 3/4" GHT(M) WZ-31307-60 Mfr # HFC241235GHT
COMPARE
Polysulfone Quick-Disconnect x GHT High-Flow Quick-Disconnect, Non-Valved PS Insert, 3/4" GHT(M)
£7.34GBP / Each
£8.81GBP (incl VAT)
CPC (Colder) HFCD13612 High-Flow Quick-Disconnect, Valved Polypropylene (PP) Body, 3/8" OD; 10/PK WZ-31308-39 Mfr # HFCD13612
COMPARE
Polypropylene Quick-Disconnect x Compression High-Flow Quick-Disconnect, Valved PP Body, 3/8" OD
£186.90GBP / Pkg of 10
£224.28GBP (incl VAT)
CPC (Colder) HFC101235GHT High-Flow Quick-Disconnect, Non-Valved Polysulfone (PS) Body, 3/4" GHT(M); 10/PK WZ-31308-76 Mfr # HFC101235GHT
COMPARE
Polysulfone Quick-Disconnect x GHT High-Flow Quick-Disconnect, Non-Valved PS Body, 3/4" GHT(M)
£154.35GBP / Pkg of 10
£185.22GBP (incl VAT)
Showing 1- 7 of 7