BriskHeat® Hazardous-Area T4-Rated Silicone Heating Blankets - 120 VAC
FM approved for Class 1 Division 2 Groups A, B, C, and D, Class II Division 2, Groups F and G
  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
These industrial-strength heating blankets are designed for use in hazardous environments. They come equipped with a high-limit safety thermostat and 1-foot long power leads extending out of a 3/4” intrinsically-safe conduit fitting. The unique grounded heating element is laminated between two layers of extra strong 23 mil and two layers of 25 mil fiberglass reinforced silicone rubber. These heaters have a power density of 2.5 watts/in2 and should only be used on metal surfaces. Approvals and compliances: FM, RoHS, REACH
Warning: Heating blankets should not be overlapped and requires properly rated temperature control and conduit.
£295.39 - £370.08GBP / Each
£354.47 - £444.10GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRX06121801T4 Hazardous T4A-Rated Silicone Heating Blanket, 6x12" , 120V, 180W WZ-36003-25 Mfr # SRX06121801T4A
COMPARE
12 158 120
£295.39GBP / Each
£354.47GBP (incl VAT)
BriskHeat SRX06243601T4 Hazardous T4A-Rated Silicone Heating Blanket, 6x24" , 120V, 360W WZ-36003-29 Mfr # SRX06243601T4A
COMPARE
24 158 120
£370.08GBP / Each
£444.10GBP (incl VAT)