BriskHeat® Hazardous-Area T3-Rated Silicone Heating Blankets with Adhesive-120 VAC
FM approved for Class 1 Division 2 Groups A, B, C, and D, Class II Division 2, Groups F and G
  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
  • easy-to-apply industrial strength pressure sensitive adhesive backing
These industrial-strength heating blankets are designed for use in hazardous environments. They come equipped with a high-limit safety thermostat and 1-foot long power leads extending out of a 3/4” intrinsically-safe conduit fitting. The unique grounded heating element is laminated between two layers of extra strong 23 mil and two layers of 25 mil fiberglass reinforced silicone rubber. These heaters have a power density of 2.5 watts/in2 and should only be used on metal surfaces. Approvals and compliances: FM, RoHS, REACH
Warning: Heating blankets should not be overlapped and requires properly rated temperature control and conduit.
£406.47 - £844.01GBP / Each
£487.76 - £1,012.81GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRX06243601PT3 Hazardous T3-Rated Silicone Heating Blanket, Adhesive, 6x24" , 120V, 360W WZ-36003-09 Mfr # SRX06243601PT3
COMPARE
24 292 120
£406.47GBP / Each
£487.76GBP (incl VAT)
BriskHeat SRX12123601PT3 Hazardous T3-Rated Silicone Heating Blanket, Adhesive, 12x12" , 120V, 360W WZ-36003-13 Mfr # SRX12123601PT3
COMPARE
12 292 120
£406.47GBP / Each
£487.76GBP (incl VAT)
BriskHeat SRX242414401PT3 Hazardous T3-Rated Silicone Heating Blanket, Adhesive, 24x24" , 120V, 1440W WZ-36003-21 Mfr # SRX242414401PT3
COMPARE
24 292 120
£844.01GBP / Each
£1,012.81GBP (incl VAT)