This product is now discontinued, View Related Products

BriskHeat® Hazardous-Area T3-Rated Silicone Heating Blankets - 120 VAC

FM approved for Class 1 Division 2 Groups A, B, C, and D, Class II Division 2, Groups F and G
  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
These industrial-strength heating blankets are designed for use in hazardous environments. They come equipped with a high-limit safety thermostat and 1-foot long power leads extending out of a 3/4” intrinsically-safe conduit fitting. The unique grounded heating element is laminated between two layers of extra strong 23 mil and two layers of 25 mil fiberglass reinforced silicone rubber. These heaters have a power density of 2.5 watts/in2 and should only be used on metal surfaces.

Approvals and compliances: FM, RoHS, REACH

Warning: Heating blankets should not be overlapped and requires properly rated temperature control and conduit.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRX06121801T3 Hazardous T3-Rated Silicone Heating Blanket, 6x12" , 120V, 180W WZ-36002-92 Mfr # SRX06121801T3
COMPARE
12 292 120
Call For Price
BriskHeat SRX06243601T3 Hazardous T3-Rated Silicone Heating Blanket, 6x24" , 120V, 360W WZ-36002-94 Mfr # SRX06243601T3
COMPARE
24 292 120
Call For Price
BriskHeat SRX12123601T3 Hazardous T3-Rated Silicone Heating Blanket, 12x12" , 120V, 360W WZ-36002-96 Mfr # SRX12123601T3
COMPARE
12 292 120
Call For Price
BriskHeat SRX12247201T3 Hazardous T3-Rated Silicone Heating Blanket, 12x24" , 120V, 720W WZ-36002-98 Mfr # SRX12247201T3
COMPARE
24 292 120
Call For Price
BriskHeat SRX242414402T3 Hazardous T3-Rated Silicone Heating Blanket, 24x24" , 120V, 1440W WZ-36003-03 Mfr # SRX242414402T3
COMPARE
24 292 240
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat